Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 1 Gallon, 12 x 24"; 100/CS

Representative image only.

Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 1 Gallon, 12 x 24"; 100/CS

Scienceware | Mfr: 13160-0009| Item: 0649730

Indicator patch darkens when sterile

₹27,200.00INR / Pkg of 100

₹27,200.00INR / Pkg of 100

Buy It Together

Current Item

Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 1 Gallon, 12 x 24"; 100/CS
Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 1 Gallon, 12 x 24"; 100/CS
0649730

₹27,200.00INR / Pkg of 100
+ =
Total Price 52200.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Precautionary procedures printed in four languages
  • Meet/exceed ASTM D 1709-85
  • Autoclavable at 250°F (121°C)

More About this Item

These high-molecular weight bags feature an indicator patch that darkens when sterile. Precautionary procedures are printed in English, French, German, and Spanish. Bags are 1.25-mil thick and meet/exceed ASTM D 1709-85.

Optional bag stands are made of aluminum tubing with baked on epoxy coating. The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand. Order stands and plastic ties separately.

Specifications
  • ColorClear
  • MaterialHDPE
  • Capacity (Gal)1
  • Length (in)24
  • Width (in)12
  • Length (cm)60.96
  • Width (cm)30.48
  • Thickness (mil)1.25
  • Description1 gal HDPE Biohazard Bags w/ Sterilization Patch, 12 x 24"; 100/cs
Reviews
Q&A