Ashcroft Model 1379 4 1/2" and 6" Process Gauges
Duragauge® allows precise pressure masurement in rough industrial environments
  • Duratube™ with as-welded-tube construction controls stress for longer life
  • Process connection constructed with “round cap tip” lowers stresses for longer life
  • Exclusive PTFE coated 400 series stainless steel rotary movement prolongs life of gauge
Type 1379 Duragauge pressure gauge is a rugged, solid-front aluminum case gauge. All cases are coated with black epoxy which withstands most environmental conditions. Gauges have 316L stainless steel wetted materials. Brass units are also available, as well as back-mount units. Contact an Applications Specialist for more details.
69,000.00 - ₹83,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft Duragauge 4.5", 0 to 160 psi 6808712 Mfr # 45-1379SSH-04L-160#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 160
₹69,000.00INR / Each
Ashcroft Duragauge 6", 0 to 1,000 psi 6808750 Mfr # 60-1379SSH-04L-1000#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 1000
₹83,100.00INR / Each