Ashcroft® Model 1008 Liquid-Filled Pressure Gauges
Liquid-filled case to dampen vibration
  • Corrosion resistant stainless steel case
  • Dry gauges are field fillable
  • PowerFlex™ movement provides superior resistance to shock, vibration and pulsation
  • True Zero™ reduces reading errors by using “zero box” instead of conventional dial pins
  • MSL helium leak tested to 1X10-6 scc/s
  • Ventable fill plug
30,300.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft 1008 Glycerin Liquid-Fill Stainless Steel Pressure Gauge, 3.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 100 psi 6842575 Mfr # 100-1008-S-L-02L-100#
COMPARE
+1.6% of span (Dry), +2% of span (Liquid-filled) 100
₹30,300.00INR / Each