Ashcroft Low Differential Pressure Transmitters
Sensor stability and repeatability provides precise differential pressure measurement
  • 4 to 20 mA output
  • Field-adjustable four-digit LCD features selectable units of in. WC, Pa, or mbar
Ashcroft model GL42 transmitters are designed for reliable and critical static or velocity pressure measurement in room pressure monitoring, building pressure, filter efficiency and fume hood monitoring. The MEMS variable-capacitance sensor provides high sensitivity and long-term stability, making these transmitters perfect for low differential pressure measurement in critical applications.
200,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Enclosure Rating Max Pressure (in WC) Min Pressure (in WC)
Availability Pricing
Ashcroft Low Differential Pressure Transmitter, 0.25 in WC 6887442 Mfr # GL42-5-MB2-42-CD-W-P25IWL
COMPARE
NEMA 4X, IP65 0.25 -0.25
₹200,000.00INR / Each
Ashcroft Low Differential Pressure Transmitter, 0.5 in WC 6887443 Mfr # GL42-5-MB2-42-CD-W-P5IWL
COMPARE
NEMA 4X, IP65 0.5 -0.5
₹200,000.00INR / Each
Ashcroft Low Differential Pressure Transmitter, 15 in WC 6887449 Mfr # GL42-5-MB2-42-CD-W-15IWL
COMPARE
NEMA 4X, IP65 15 -15
₹200,000.00INR / Each