Ashcroft® Commercial Gauges with Bottom Connection
Gauges feature Powerflex™ shock and vibration protection
  • True zero indication identifies damaged gauges — increases safety and ensures accuracy
Use these glycerin-filled gauges for easier reading when high-frequency pressure fluctuations or vibrations are present. Gauges construction includes brass socket, phosphor bronze tube, 304 stainless steel case, and plastic lens. Bottom and back connections are 1/4" NPT(M).
10,400.00 - ₹11,700.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Accuracy Description
Availability Pricing
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 30 to 0" Hg, 2-1/2" Dial 0738063 Mfr # 63-W-1008-A-L-02L-30IHGV
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 30 to 0" Hg, 2-1/2" Dial
₹10,600.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 15 psi, 2-1/2" Dial 0738065 Mfr # 63-1008-A-L-02L-15#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 15 psi, 2-1/2" Dial
₹10,600.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 30 psi, 2-1/2" Dial 0738070 Mfr # 63-W-1008-A-L-02L-30#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 30 psi, 2-1/2" Dial
₹10,400.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 60 psi, 2-1/2" Dial 0738075 Mfr # 63-W-1008-A-L-02L-60#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 60 psi, 2-1/2" Dial
₹10,400.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 100 psi, 2-1/2" Dial 0738080 Mfr # 63-W-1008-A-L-02L-100#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 100 psi, 2-1/2" Dial
₹10,400.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 160 psi, 2-1/2" Dial 0738085 Mfr # 63-W-1008-A-L-02L-160#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 160 psi, 2-1/2" Dial
₹10,400.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 200 psi, 2-1/2" Dial 0738090 Mfr # 63-W-1008-A-L-02L-200#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 200 psi, 2-1/2" Dial
₹10,400.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 1000 psi, 2-1/2" Dial 0738092 Mfr # 63-W-1008-A-L-02L-1000#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 1000 psi, 2-1/2" Dial
₹11,700.00INR / Each
Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 0 to 3000 psi, 2 1/2" Dial 0738094 Mfr # 63-W-1008-A-L-02L-3000#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) Commercial Gauge with Bottom Connection, 0 to 3000 psi, 2 1/2" Dial
₹11,700.00INR / Each
Showing 1- 9 of 9