Cole-Parmer® Dot- and Strip-Style Cryogenic Labels
No need for separate labels—work in cryogenic applications, ultra-low freezers, and incubators
  • Inert polyester with a unique adhesive adheres to glass, plasticware, and most metals
  • No need to self-laminate and sleeker labels fit most storage systems without adding bulk—even cryocanes
  • Strips and dots come in varying sizes, colors, and on rolls for individual labeling or on sheets for use with laser printers
These labels withstand conventional freezing (-80 °C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196 °C). Choose the size, color, and format to suit your labeling needs. The wide range of sizes allow you to label your whole experiment permanently. From microcentrifuge tube to cryovial; microtiter plate to storage box.
5,800.00 - ₹44,800.00INR / Pkg of 3840
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Label Dots, Laser Printer, 9.5 mm dia, White; 192 x 20 Sheets
₹22,600.00INR / Pkg of 3840
COMPARE
Label Dots, 13 mm dia, White; 1000/Pk
₹7,400.00INR / Pkg of 1000
COMPARE
Label Strips, 33 x 13 mm, White; 1000/Pk
₹14,700.00INR / Pkg of 1000
COMPARE
Label Strips, 20 x 6 mm, White; 1000/Pk
₹6,800.00INR / Pkg of 1000
COMPARE
Label Dots, 9.5 mm dia, White; 1000/Pk
₹6,800.00INR / Pkg of 1000
COMPARE
Label Dots, 9.5 mm dia, White; 5000/Pk
₹30,200.00INR / Pkg of 5000
COMPARE
Label Dots, 19 mm Dia, White; 5000/Pk
₹44,800.00INR / Pkg of 5000
COMPARE
Label Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets
₹28,500.00INR / Pkg of 2160
COMPARE
Label Strips, Laser Printer, 33 x 13 mm, White; 85 x 20 Sheets
₹15,200.00INR / Pkg of 1700
COMPARE
Label Strips, Laser Printer, 33 x 13 mm, Multi; 85 x 25 Sheets
₹18,700.00INR / Pkg of 2125
COMPARE
Label Strips, Laser Printer, 24 x 13 mm, White; 119 x 20 Sheets
₹14,600.00INR / Pkg of 2380
COMPARE
Label Strips, Laser Printer, 67 x 25 mm, White; 30 x 20 Sheets
₹25,800.00INR / Pkg of 600
COMPARE
Label Strips, Laser Printer, 38 x 6 mm, White; 156 x 20 Sheets
₹19,600.00INR / Pkg of 3120
COMPARE
Label Strips, 33 x 13 mm, Blue; 1000/Roll
₹14,000.00INR / Roll of 1000
COMPARE
Label Strips, 33 x 13 mm, Red; 1000/Roll
₹14,000.00INR / Roll of 1000
COMPARE
Label Strips, 33 x 13 mm, Green; 1000/Roll
₹8,000.00INR / Roll of 1000
COMPARE
Label Strips, 33 x 13 mm, Yellow; 1000/Roll
₹8,000.00INR / Roll of 1000
COMPARE
Label Strips, 33 x 13 mm, Purple; 1000/Roll
₹14,000.00INR / Roll of 1000
COMPARE
Label Dots, 9.5 mm dia, Multi-Color; 5000/Roll
₹29,200.00INR / Roll of 5000
COMPARE
Label Dots, 9.5 mm dia, Blue; 1000/Roll
₹6,800.00INR / Roll of 1000
Showing 1 - 20 of 24