Cole-Parmer Label Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets
Representative image only.
Cole-Parmer Label Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets
Cole-Parmer | Item: 0439672
No need for separate labels—work in cryogenic applications, ultra-low freezers, and incubators
₹28,500.00INR / Pkg of 2160
₹28,500.00INR / Pkg of 2160
Buy It Together
Current Item
Cole-Parmer Label Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets
Cole-Parmer Label Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets
0439672
₹28,500.00INR / Pkg of 2160
+ +
Tough-Spots 9185-0000 Small Labels, White, 3/8" Dia; 1000/RL
6220700
₹9,000.00INR / Roll of 1000
=
Total Price 55200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Inert polyester with a unique adhesive adheres to glass, plasticware, and most metals
  • No need to self-laminate and sleeker labels fit most storage systems without adding bulk—even cryocanes
  • Strips and dots come in varying sizes, colors, and on rolls for individual labeling or on sheets for use with laser printers
More About this Item
These labels withstand conventional freezing (-80 °C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196 °C). Choose the size, color, and format to suit your labeling needs. The wide range of sizes allow you to label your whole experiment permanently. From microcentrifuge tube to cryovial; microtiter plate to storage box.
Dots are suitable for use on cryotubes, glass vials, and microcentrifuge tubes.
Specifications
  • First color changeWhite
  • Diameter (in)3/4
  • DescriptionLabel Dots, Laser Printer, 19 mm dia, White; 108 x 20 Sheets