Ansell TouchNTuff® 92-675 Disposable Nitrile Gloves
Robust gloves offer excellent chemical splash protection
  • Nitrile is a durable and soft material for strength and comfort
  • Get a secure grip with textured fingertips
  • Complies with FDA food handling requirements
TouchNTuff disposable powder-free nitrile gloves are appropriate for transferring liquids or solids, sample taking/processing, handling of incoming goods, maintenance, and food processing. Textured fingertips enhance wet or dry grip. Ambidextrous with beaded cuffs. Blue color.
3,500.00 - ₹46,100.00INR / Box of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Grip Material Size
Availability Pricing
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size S (6.5-7); 100/Box 8160196 Mfr # 105081 BOX
COMPARE
Textured Nitrile Small
₹4,100.00INR / Box of 100
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size M (7.5-8); 100/Box 8160197 Mfr # 105082 BOX
COMPARE
Textured Nitrile Medium
₹3,700.00INR / Box of 100
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size L (8.5-9); 100/Box 8160198 Mfr # 105083 BOX
COMPARE
Textured Nitrile Large
₹3,800.00INR / Box of 100
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size XL (9.5-10); 100/Box 8160199 Mfr # 105084 BOX
COMPARE
Textured Nitrile X-Large
₹3,500.00INR / Box of 100
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size S (6.5-7); Case of 1000 (10 boxes of 100 gloves) 8160276 Mfr # 105081
COMPARE
Textured Nitrile Small
₹37,000.00INR / Case of 1000
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size L (8.5-9); Case of 1000 (10 boxes of 100 gloves) 8160278 Mfr # 105083
COMPARE
Textured Nitrile Large
₹46,100.00INR / Case of 1000
Ansell TouchNTuff® 92-675 Nitrile Gloves with Textured Fingertips, Size XL (9.5-10); 1000/CS (10 boxes of 100 gloves) 8160279 Mfr # 105084
COMPARE
Textured Nitrile X-Large
₹37,000.00INR / Case of 1000
Showing 1- 7 of 7