Analtech Glass-Backed Preparative TLC Uniplates

Efficient separations — economically!
  • Constructed of glass backing
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect preparative plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
87,300.00 - ₹146,000.00INR / Pkg of 25

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Layer Thickness (mm) Plate Length (cm) Plate Width (cm)
Availability Pricing
Analtech Preparative Uniplates, 20x20cm/Reversed/500mm Thick/15um Particle; 25/Pk 3410270 Mfr # P50012
COMPARE
500 20 20
₹146,000.00INR / Pkg of 25
Analtech Preparative Uniplates, 20x20cm/Silica G/1000mm Thick/15um Part; 25/Pk 3410300 Mfr # P01013
COMPARE
1000 20 20
₹87,300.00INR / Pkg of 25
Analtech Preparative Uniplates, 20x20cm/UV/Silica G/1000mm Thick/15um Part; 25/Pk 3410305 Mfr # P02013
COMPARE
1000 20 20
₹87,300.00INR / Pkg of 25
Analtech Preparative Uniplates, 20x20cm/UV/Silica G/2000mm Thick/15um Part; 25/Pk 3410315 Mfr # P02015
COMPARE
2000 20 20
₹130,000.00INR / Pkg of 25
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat