AEMC Clamp-On Ground Resistance Testers
Units display touch voltage alerting of potential unsafe conditions
  • Test frequency selection increases accuracy in inductive environments
  • Touch voltage indication sounds a warning buzzer and displays an icon to caution you of dangerous conditions
  • Loop resistance function alerts that measurement may not be ground resistance for added safety
  • Model 6417 displays stored measurements on the display or wirelessly, up to 30 feet (9 meters), by Bluetooth® connectivity to PC or Android™ based mobile application
These ground resistance testers measure ground rod, grid resistance, and impedance without using auxiliary rods and are designed for use in multi-grounded systems without disconnecting or de-energizing the ground system under test. Testers are also used to measure resistance and continuity of grounding loops around pads and buildings. High-sensitivity measurement function enables measurement of leakage current flowing to ground or circulating in ground loops from 0.2 mA to 40 A and resistances from 0.01 to 1500.
Model 6417 includes PC based DataView softwared providing the ability to configure and retrieve data through Bluetooth communication and report generation capability. Use the Android™ based app with model 6417 for configuring, downloading, printing and emailing test results from your tablet or smartphone. The GPS capability in your device provides the ability to easily locate the site associated with the measurements.
443,000.00 - ₹520,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
AEMC 6416 Ground Resistance Tester with Alarm and Memory 2004748 Mfr # 2141.01
COMPARE
₹443,000.00INR / Each
AEMC 6417 Ground Resistance Tester with Bluetooth, Alarm and Memory 2004749 Mfr # 2141.02
COMPARE
₹520,000.00INR / Each