Advantec Stainless Steel Pressure Filtration Holders
Stainless steel type 304 or 316 minimizes resistance and maximizes flow rate
  • Holders are autoclavable with one filter membrane that is sealed with removable wing nuts
  • Vent to release pressure
Pressure filtration holders maximize flow rates by applying positive pressure to the fluid over a broad membrane surface. Holders have PTFE gaskets, silicone O-rings, and PTFE-coated photoetched stainless steel support screens (model 02926-50 has a 304 SS support screen) to prevent nitrocellulose membranes from sticking. Holders process samples from an in-line system or pressure vessel.
910,000.00 - ₹1,488,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Filter Diameter (mm) Material
Availability Pricing
Advantec 300800 Pressure Filtration Holder, for 90 mm filter; 304 Stainless Steel 0292650 Mfr # 300800
COMPARE
90 304 Stainless Steel
₹910,000.00INR / Each
Advantec 301900 Pressure Filtration Holder, for 142 mm filter; 304 Stainless Steel 0292750 Mfr # 301900
COMPARE
142 304 Stainless Steel
₹1,488,000.00INR / Each
Advantec 302000 Pressure Filtration Holder for 142 mm filters; 316 Stainless Steel 2990002 Mfr # 302000
COMPARE
142 316 Stainless Steel
₹1,447,000.00INR / Each