Advantec Stainless Steel Pressure Filtration Holders with Reservoir
Stainless steel type 304 or 316 minimizes resistance and maximizes flow rate
Pressure filtration holders maximize flow rates by applying positive pressure to the fluid over a broad membrane surface. Holders are autoclavable with one filter membrane that is sealed with removable wing nuts. All holders vent to release pressure.
Holders have PTFE gaskets, silicone O-rings, and PTFE-coated photoetched stainless steel support screens to prevent nitrocellulose membranes from sticking.
Stainless Steel Holders with Reservoir let you filter samples without a separate pressure-dispensing vessel. The 47-mm holder has a 200-mL capacity, the 90-mm holder has a 750-mL capacity, and the 142-mm holders have a 1500-mL capacity.
826,000.00 - ₹2,212,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Filter Diameter (mm) Material
Availability Pricing
Advantec 301500 304 SS Pressure Filtration Holder w/ Reservoir, 47 mm 0292632 Mfr # 301500
COMPARE
47 304 Stainless Steel
₹1,137,000.00INR / Each
Advantec 361600 304 SS Pressure Filtration Holder w/ Reservoir, 90 mm 0292654 Mfr # 361600
COMPARE
90 304 Stainless Steel
₹826,000.00INR / Each
Advantec 302300 304 SS Pressure Filtration Holder w/ Reservoir, 142 mm 0292700 Mfr # 302300
COMPARE
142 304 Stainless Steel
₹2,212,000.00INR / Each
Advantec 302400 316 SS Pressure Filtration Holder w/ Reservoir, 142 mm 0293300 Mfr # 302400
COMPARE
142 316 Stainless Steel