Advantec PTFE Hydrophilic Membranes
Hydrophilic PTFE membranes offer maximum chemical and pH resistance with minimal aqueous extractables (<0.3 wt%). Since the filter is optically clear when wet it is ideally suited for culture and microscopic examination of cells/particles captured without requiring a separate step. Maximum operating temperature is 100°C (212°F). Autoclaving and allowing filter to dry will render it hydrophobic.
58,300.00 - ₹127,000.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm)
Availability Pricing
Advantec H010A025A PTFE Hydrophilic Membranes; 25 mm dia; 0.10 µm; 100/PK 3622952 Mfr # H010A025A
COMPARE
25 0.1
₹67,300.00INR / Pkg of 100
Advantec H010A047A PTFE Hydrophilic Membranes; 47 mm dia; 0.10 µm; 100/PK 3622954 Mfr # H010A047A
COMPARE
47 0.1
₹91,400.00INR / Pkg of 100
Advantec H010A090C PTFE Hydrophilic Membranes; 90 mm dia; 0.10 µm; 25/PK 3622956 Mfr # H010A090C
COMPARE
90 0.1
₹74,500.00INR / Pkg of 25
Advantec H050A047A PTFE Hydrophilic Membranes; 47 mm dia; 0.50 µm; 100/PK 3622962 Mfr # H050A047A
COMPARE
47 0.5
₹91,400.00INR / Pkg of 100
Advantec H050A090C PTFE Hydrophilic Membranes; 90 mm dia; 0.50 µm; 25/PK 3622964 Mfr # H050A090C
COMPARE
90 0.5
₹74,500.00INR / Pkg of 25
Advantec H100A047A PTFE Hydrophilic Membranes; 47 mm dia; 1.00 µm; 100/PK 3622970 Mfr # H100A047A
COMPARE
47 1
₹91,400.00INR / Pkg of 100
Advantec H100A090C PTFE Hydrophilic Membranes; 90 mm dia; 1.00 µm; 25/PK 3622972 Mfr # H100A090C
COMPARE
90 1
₹74,500.00INR / Pkg of 25
Advantec H020A013A PTFE Hydrophilic Membranes; 13 mm dia; 0.20 µm; 100/PK 3632940 Mfr # H020A013A
COMPARE
13 0.2
₹58,300.00INR / Pkg of 100
Advantec H020A025A PTFE Hydrophilic Membranes; 25 mm dia; 0.20 µm; 100/PK 3632942 Mfr # H020A025A
COMPARE
25 0.2
₹67,300.00INR / Pkg of 100
Advantec H020A047A PTFE Hydrophilic Membranes; 47 mm dia; 0.20 µm; 100/PK 3632944 Mfr # H020A047A
COMPARE
47 0.2
₹91,400.00INR / Pkg of 100
Advantec H020A090C PTFE Hydrophilic Membranes; 90 mm dia; 0.20 µm; 25/PK 3632946 Mfr # H020A090C
COMPARE
90 0.2
₹74,500.00INR / Pkg of 25
Advantec H010A142C PTFE Hydrophilic Membranes; 142 mm dia; 0.10 µm; 25/PK 3632947 Mfr # H010A142C
COMPARE
142 0.1
₹127,000.00INR / Pkg of 25
Advantec H020A142C PTFE Hydrophilic Membranes; 142 mm dia; 0.20 µm; 25/PK 3632948 Mfr # H020A142C
COMPARE
142 0.2
₹109,000.00INR / Pkg of 25
Advantec H050A142C PTFE Hydrophilic Membranes; 142 mm dia; 0.50 µm; 25/PK 3632949 Mfr # H050A142C
COMPARE
142 0.5
₹109,000.00INR / Pkg of 25
Showing 1- 14 of 14