Advantec PTFE Depth Filters
Ideal for aggressive chemicals and air filtration applications
  • High temperature and strong chemical resistance
  • Excellent air permeability with minimal pressure drop
  • High loading capacity
Pure PTFE fibers are sintered to improve handling characteristics and minimize fiber slough-off and downstream contamination. Recommended for filtering hot acids, strong solvents, and for applications where media contamination must be at a critically low level. Use these hydrophobic filters to separate aqueous and non-aqueous phases of mixtures as well as for air/gas filtration or venting. Temperature range is –120 to 260°C (–184 to 436°F).
7,900.00 - ₹50,800.00INR / Pkg of 10
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Pore Size (micron) Description
Availability Pricing
Advantec PTFE Depth Filter, 2.0um pore, 150-mm diameter; pack of 5 3624001 Mfr # PF020150MM
COMPARE
PTFE 2 PTFE Depth Filter, 2.0um pore, 150-mm diameter; pack of 5
₹50,800.00INR / Pkg of 5
Advantec PTFE Depth Filter, 2 µm pore, 47-mm diameter; pack of 10 3624002 Mfr # PF02047MM
COMPARE
PTFE 2 PTFE Depth Filter, 2 µm pore, 47-mm diameter; pack of 10
₹13,800.00INR / Pkg of 10
Advantec PTFE Depth Filter, 4um pore, 47-mm diameter; pack of 10 3624004 Mfr # PF04047MM
COMPARE
PTFE 4 PTFE Depth Filter, 4um pore, 47-mm diameter; pack of 10
₹13,800.00INR / Pkg of 10
Advantec PTFE Depth Filter, 5 µm pore, 47-mm diameter; pack of 20 3624006 Mfr # PF05047MM
COMPARE
PTFE 5 PTFE Depth Filter, 5 µm pore, 47-mm diameter; pack of 20
₹11,000.00INR / Pkg of 20
Advantec PTFE Depth Filter, 10 µm pore, 47-mm diameter; pack of 20 3624008 Mfr # PF10047MM
COMPARE
PTFE 10 PTFE Depth Filter, 10 µm pore, 47-mm diameter; pack of 20
₹11,000.00INR / Pkg of 20
Advantec PTFE Depth Filter, 6 µm pore, 47-mm diameter; pack of 20 3624009 Mfr # PF06047MM
COMPARE
PTFE 6 PTFE Depth Filter, 6 µm pore, 47-mm diameter; pack of 20
₹18,800.00INR / Pkg of 20
Advantec PTFE Depth Filter, 2 µm pore, 25-mm diameter; pack of 10 3624012 Mfr # PF02025MM
COMPARE
PTFE 2 PTFE Depth Filter, 2 µm pore, 25-mm diameter; pack of 10
₹13,800.00INR / Pkg of 10
Advantec PTFE Depth Filter, 5 µm pore, 25-mm diameter; pack of 20 3624016 Mfr # PF05025MM
COMPARE
PTFE 5 PTFE Depth Filter, 5 µm pore, 25-mm diameter; pack of 20
₹7,900.00INR / Pkg of 20
Advantec PTFE Depth Filter, 10 µm pore, 25-mm diameter; pack of 20 3624018 Mfr # PF10025MM
COMPARE
PTFE 10 PTFE Depth Filter, 10 µm pore, 25-mm diameter; pack of 20
₹10,600.00INR / Pkg of 20
Advantec PTFE Depth Filter, 2 µm pore, 55-mm diameter; pack of 10 3624022 Mfr # PF02055MM
COMPARE
PTFE 2 PTFE Depth Filter, 2 µm pore, 55-mm diameter; pack of 10
₹16,700.00INR / Pkg of 10
Advantec PTFE Depth Filter, 5 µm pore, 55-mm diameter; pack of 20 3624026 Mfr # PF05055MM
COMPARE
PTFE 5 PTFE Depth Filter, 5 µm pore, 55-mm diameter; pack of 20
₹13,400.00INR / Pkg of 20
Advantec PTFE Depth Filter, 6 µm pore, 55-mm diameter; pack of 20 3624029 Mfr # PF06055MM
COMPARE
PTFE 6 PTFE Depth Filter, 6 µm pore, 55-mm diameter; pack of 20
₹22,700.00INR / Pkg of 20
Advantec PTFE Depth Filter, 5 µm pore, 8" x 10" sheet (20 x 25 cm); pack of 5 3624036 Mfr # PF0508X10IN
COMPARE
PTFE 5 PTFE Depth Filter, 5 µm pore, 8" x 10" sheet (20 x 25 cm); pack of 5
₹33,000.00INR / Pkg of 5
Advantec PF100 Depth Filter, 10 um PTFE,  8" x 10" sheet (20 x 25 cm); pack of 5 3624038 Mfr # PF1008X10IN
COMPARE
PTFE 10 PF100 Depth Filter, 10 um PTFE, 8" x 10" sheet (20 x 25 cm); pack of 5
₹33,000.00INR / Pkg of 5
Showing 1- 14 of 14