Advantec Pressure Vessels
Wide-mouth opening allows easy cleaning
  • Pressure vessels are ASME certified
Use these electropolished stainless steel pressure vessels to propel filtration fluids through pressure holders by applying external pressure from compressed gases. Increasing pressure increases the efficiency of the EPR (ethylene propylene rubber) O-ring seal.
Vessels feature 5-7⁄8"x 4-7⁄8" (14.9 x 12.3 cm) wide-mouth opening for easy cleaning, SS dip tube, and molded neoprene rubber base for stability. Intake for the dip tube, located at the deepest point in the vessel, minimizing fluid loss. Sterilize vessels at 250°F (121°C) for 30 minutes. Make connections to four 1⁄4" NPT(F) ports.
610,000.00 - ₹1,765,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Material
Availability Pricing
Advantec 740140 Pressure Vessel; 304 SS; 1 gal. (3.78 L) capacity; Wide-Mouth 2990200 Mfr # 740140
COMPARE
1 304 Stainless Steel
₹679,000.00INR / Each
Advantec 740340 Pressure Vessel; 304 SS; 3 gal. (11.3 L) capacity; Wide-Mouth 2990202 Mfr # 740340
COMPARE
3 304 Stainless Steel
₹727,000.00INR / Each
Advantec 740540 Pressure Vessel; 304 SS; 5 gal. (18.9 L) capacity; Wide-Mouth 2990204 Mfr # 740540
COMPARE
5 304 Stainless Steel
₹786,000.00INR / Each
Advantec 740160 Pressure Vessel; 316 SS; 1 gal. (3.78 L) capacity; Wide-Mouth 2990290 Mfr # 740160
COMPARE
1 316 Stainless Steel
₹610,000.00INR / Each
Advantec 740360 Pressure Vessel; 316 SS; 3 gal. (11.3 L) capacity; Wide-Mouth 2990292 Mfr # 740360
COMPARE
3 316 Stainless Steel
₹637,000.00INR / Each
Advantec 740560 Pressure Vessel; 316 SS; 5 gal. (18.9 L) capacity; Wide-Mouth 2990294 Mfr # 740560
COMPARE
5 316 Stainless Steel
₹679,000.00INR / Each
Advantec 740860 Pressure Vessel; 316 SS; 8 gal. (30.2 L) capacity; Wide-Mouth 2990298 Mfr # 740860
COMPARE
8 316 Stainless Steel
₹1,765,000.00INR / Each
Showing 1- 7 of 7