Advantec Milk Sediment Discs
Smaller retention offers better results than lintine paper
Use these refined cellulose fiber filters for determining the amount of sediment or insoluble material in raw milk and other dairy products. Available in white or black color; the black disc allows easier identification of white particulates.
8,300.00 - ₹16,400.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec N01026 Milk Sediment Discs, White, 33MM 50/PK 0664597 Mfr # N01026
COMPARE
Milk Sediment Discs, White, 33MM 50/PK
₹8,300.00INR / Pkg of 50
Advantec N01026-B Milk Sediment Discs, Black, 33MM 50/PK 0664698 Mfr # N01026-B
COMPARE
Milk Sediment Discs, Black, 33MM 50/PK
₹16,400.00INR / Pkg of 50