Advantec® Hydrophobic PTFE Membranes with Support
Ideal for air and gas venting
  • Protects vacuum pumps and critical samples by blocking aqueous liquids
  • Operating temperature range is –31 to 266°F (–35 to 130°C)
A laminated polypropylene support offers improved handling characteristics to this PTFE membrane filter. Use to sterilize and clarify strong acids, bases, and aggressive solvents.
65,000.00 - ₹162,000.00INR / Pkg of 100
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Advantec J020A025A PTFE Hydrophobic Membranes with Support; 25 mm dia; 0.20 µm; 100/PK 0291652 Mfr # J020A025A
COMPARE
25 Hydrophobic PTFE 0.2
₹65,000.00INR / Pkg of 100
Advantec J050A025A PTFE Hydrophobic Membranes with Support; 25 mm dia; 0.50 µm; 100/PK 0291653 Mfr # J050A025A
COMPARE
25 Hydrophobic PTFE 0.5
₹69,600.00INR / Pkg of 100
Advantec J020A047A PTFE Hydrophobic Membranes with Support; 47 mm dia; 0.20 µm; 100/PK 0291654 Mfr # J020A047A
COMPARE
47 Hydrophobic PTFE 0.2
₹91,600.00INR / Pkg of 100
Advantec J020A090C PTFE Hydrophobic Membranes with Support; 90 mm dia; 0.20 µm; 25/PK 0291656 Mfr # J020A090C
COMPARE
90 Hydrophobic PTFE 0.2
₹73,800.00INR / Pkg of 25
Advantec J020A142C PTFE Hydrophobic Membranes with Support; 142 mm dia; 0.20 µm; 25/PK 0291658 Mfr # J020A142C
COMPARE
142 Hydrophobic PTFE 0.2
₹114,000.00INR / Pkg of 25
Advantec J050A047A PTFE Hydrophobic Membranes with Support; 47 mm dia; 0.50 µm; 100/PK 0291664 Mfr # J050A047A
COMPARE
47 Hydrophobic PTFE 0.5
₹90,700.00INR / Pkg of 100
Advantec J050A142C PTFE Hydrophobic Membranes with Support; 142 mm dia; 0.50 µm; 25/PK 0291668 Mfr # J050A142C
COMPARE
142 Hydrophobic PTFE 0.5
₹114,000.00INR / Pkg of 25
Advantec J100A025A PTFE Hydrophobic Membranes with Support; 25 mm dia; 1.00 µm; 100/PK 0291672 Mfr # J100A025A
COMPARE
25 Hydrophobic PTFE 1
₹69,600.00INR / Pkg of 100
Advantec J100A047A PTFE Hydrophobic Membranes with Support; 47 mm dia; 1.00 µm; 100/PK 0291674 Mfr # J100A047A
COMPARE
47 Hydrophobic PTFE 1
₹90,700.00INR / Pkg of 100
Advantec J010A047A PTFE Hydrophobic Membranes with Support; 47 mm dia; 0.10 µm; 100/PK 3632964 Mfr # J010A047A
COMPARE
47 Hydrophobic PTFE 0.1
₹93,400.00INR / Pkg of 100
Advantec J050A293D PTFE Hydrophobic Membranes with Support; 293 mm dia; 0.50 µm; 10/PK 3632972 Mfr # J050A293D
COMPARE
293 Hydrophobic PTFE 0.5
₹162,000.00INR / Pkg of 10
Showing 1- 11 of 11