Advantec Grade No. 7 Ashless Filter Paper for Medium Retention
Manufactured to exacting standards for uniform performance
  • Highest purity for retaining medium particles (5 to 10 m), precise gravimetric analysis
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Sold in packs of 100.
4,600.00 - ₹17,800.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N077.0CM Grade No. 7 Ashless Quantitative Filter Paper; 7.0 cm dia.; pack of 100 8105178 Mfr # N077.0CM
COMPARE
7 Alpha Cotton Cellulose
₹4,600.00INR / Pkg of 100
Advantec N0718.5CM Grade 7 Ashless Quantitative Filters, 5-10 um, 18.5 cm; 100/Pk 8105188 Mfr # N0718.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹17,800.00INR / Pkg of 100