Advantec Grade No. 6 Ashless Filter Paper for Medium-Fine Retention
Manufactured to exacting standards for uniform performance
  • Retains medium-fine particulates (2 to 10 m), trace and precious metals
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
5,400.00 - ₹17,200.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N069.0CM Grade No. 6 Ashless Quantitative Filter Paper; 9.0 cm dia.; pack of 100 8105160 Mfr # N069.0CM
COMPARE
9 Alpha Cotton Cellulose
₹5,400.00INR / Pkg of 100
Advantec N0612.5CM Grade 6 Ashless Quantitative Filters, 2-10 um, 12.5 cm; 100/Pk 8105170 Mfr # N0612.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹6,600.00INR / Pkg of 100
Advantec N0615.0CM Grade 6 Ashless Quantitative Filters, 2-10 um, 15 cm; 100/Pk 8105172 Mfr # N0615.0CM
COMPARE
15 Alpha Cotton Cellulose
₹8,400.00INR / Pkg of 100
Advantec N0618.5CM Grade 6 Ashless Quantitative Filters, 2-10 um, 18.5 cm; 100/Pk 8105174 Mfr # N0618.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹17,200.00INR / Pkg of 100