Advantec Grade No. 5C Ashless Filter Paper for Fine Retention
Manufactured to exacting standards for uniform performance
  • Collect fine precipitates (<5 m) such as SrSO4, BaSO4, HgCrO4, and colloidal dispersions; gravimetric analysis
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
3,800.00 - ₹17,400.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N05C5.5CM Grade 5C Ashless Quantitative Filters, <5 um, 5.5 cm; 100/Pk 8105142 Mfr # N05C5.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹3,800.00INR / Pkg of 100
Advantec N05C7.0CM Grade 5C Ashless Quantitative Filters, <5 um, 7 cm; 100/Pk 8105144 Mfr # N05C7.0CM
COMPARE
7 Alpha Cotton Cellulose
₹4,100.00INR / Pkg of 100
Advantec N05C9.0CM Grade 5C Ashless Quantitative Filters, <5 um, 9 cm; 100/Pk 8105146 Mfr # N05C9.0CM
COMPARE
9 Alpha Cotton Cellulose
₹5,300.00INR / Pkg of 100
Advantec N05C11.0CM Grade 5C Ashless Quantitative Filters, <5 um, 11 cm; 100/Pk 8105148 Mfr # N05C11.0CM
COMPARE
11 Alpha Cotton Cellulose
₹7,200.00INR / Pkg of 100
Advantec N05C12.5CM Grade 5C Ashless Quantitative Filters, <5 um, 12.5 cm; 100/Pk 8105150 Mfr # N05C12.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹8,800.00INR / Pkg of 100
Advantec N05C15.0CM Grade 5C Ashless Quantitative Filters, <5 um, 15 cm; 100/Pk 8105152 Mfr # N05C15.0CM
COMPARE
15 Alpha Cotton Cellulose
₹11,200.00INR / Pkg of 100
Advantec N05C18.5CM Grade 5C Ashless Quantitative Filters, <5 um, 18.5 cm; 100/Pk 8105154 Mfr # N05C18.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹17,400.00INR / Pkg of 100
Showing 1- 7 of 7