Advantec Grade No. 5B Ashless Filter Paper for Medium Retention
Manufactured to exacting standards for uniform performance
  • Retains medium particles (5 to 10 m) such as CaCO3, PbSO4, CaCO4, MnCO3, ZnCO3, ZnS, AgCl
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
3,800.00 - ₹17,500.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N05B5.5CM Grade 5B Ashless Quantitative Filters, 5-10 um, 5.5 cm; 100/Pk 8105112 Mfr # N05B5.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹3,800.00INR / Pkg of 100
Advantec N05B7.0CM Grade 5B Ashless Quantitative Filters, 5-10 um, 7 cm; 100/Pk 8105114 Mfr # N05B7.0CM
COMPARE
7 Alpha Cotton Cellulose
₹4,300.00INR / Pkg of 100
Advantec N05B9.0CM Grade 5B Ashless Quantitative Filters, 5-10 um, 9 cm; 100/Pk 8105116 Mfr # N05B9.0CM
COMPARE
9 Alpha Cotton Cellulose
₹5,500.00INR / Pkg of 100
Advantec N05B11.0CM Grade 5B Ashless Quantitative Filters, 5-10 um, 11 cm; 100/Pk 8105118 Mfr # N05B11.0CM
COMPARE
11 Alpha Cotton Cellulose
₹7,300.00INR / Pkg of 100
Advantec N05B12.5CM Grade 5B Ashless Quantitative Filters, 5-10 um, 12.5 cm; 100/Pk 8105120 Mfr # N05B12.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹8,800.00INR / Pkg of 100
Advantec N05B15.0CM Grade 5B Ashless Quantitative Filters, 5-10 um, 15 cm; 100/Pk 8105122 Mfr # N05B15.0CM
COMPARE
15 Alpha Cotton Cellulose
₹11,400.00INR / Pkg of 100
Advantec N05B18.5CM Grade 5B Ashless Quantitative Filters, 5-10 um, 18.5 cm; 100/Pk 8105124 Mfr # N05B18.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹17,500.00INR / Pkg of 100
Showing 1- 7 of 7