Advantec Grade No. 4A Hardened Ashless Filter Paper for Very Fine Retention

Manufactured to exacting standards for uniform performance
  • High wet strength, suitable for use under high pressure, high chemical and pH resistance, retains fine crystalline precipitates (<5 m), slow flow
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Sold in packs of 100.
5,300.00 - ₹11,500.00INR / Pkg of 100

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N04A9.0CM Grade 4A Ashless Quantitative Filters, <5 um, 9 cm; 100/Pk 8105194 Mfr # N04A9.0CM
COMPARE
9 Alpha Cotton Cellulose
₹5,300.00INR / Pkg of 100
Advantec N04A11.0CM Grade 4A Ashless Quantitative Filters, <5 um, 11 cm; 100/Pk 8105196 Mfr # N04A11.0CM
COMPARE
11 Alpha Cotton Cellulose
₹6,800.00INR / Pkg of 100
Advantec N04A12.5CM Grade 4A Ashless Quantitative Filters, <5 um, 12.5 cm; 100/Pk 8105198 Mfr # N04A12.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹8,300.00INR / Pkg of 100
Advantec N04A15.0CM Grade 4A Ashless Quantitative Filters, <5 um, 15 cm; 100/Pk 8105200 Mfr # N04A15.0CM
COMPARE
15 Alpha Cotton Cellulose
₹11,500.00INR / Pkg of 100
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat