Advantec Grade No. 3 Ashless Filter Paper for Medium Retention
Manufactured to exacting standards for uniform performance
  • Medium retention (5 to 10 m), fast flow rate analysis of soils, fertilizers, cement, and minerals
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
3,100.00 - ₹12,400.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N035.5CMM Grade No. 3 Ashless Quantitative Filter Paper; 5.5 cm dia.; pack of 100 8105056 Mfr # N035.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹3,100.00INR / Pkg of 100
Advantec N037.0CMM Grade No. 3 Ashless Quantitative Filter Paper; 7.0 cm dia.; pack of 100 8105058 Mfr # N037.0CM
COMPARE
7 Alpha Cotton Cellulose
₹4,400.00INR / Pkg of 100
Advantec N039.0CMCM Grade No. 3 Ashless Quantitative Filter Paper; 9.0 cm dia.; pack of 100 8105060 Mfr # N039.0CM
COMPARE
9 Alpha Cotton Cellulose
₹5,300.00INR / Pkg of 100
Advantec N0311.0CM Grade 3 Ashless Quantitative Filters, 5-10 um, 11 cm; 100/Pk 8105062 Mfr # N0311.0CM
COMPARE
11 Alpha Cotton Cellulose
₹6,600.00INR / Pkg of 100
Advantec N0312.5CM Grade 3 Ashless Quantitative Filters, 5-10 um, 12.5 cm; 100/Pk 8105064 Mfr # N0312.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹8,300.00INR / Pkg of 100
Advantec N0315.0CM Grade 3 Ashless Quantitative Filters, 5-10 um, 15 cm; 100/Pk 8105066 Mfr # N0315.0CM
COMPARE
15 Alpha Cotton Cellulose
₹10,900.00INR / Pkg of 100
Advantec N0318.5CMM Grade 3 Ashless Quantitative Filters, 5-10 um, 18.5 cm; 100/Pk 8105068 Mfr # N0318.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹12,400.00INR / Pkg of 100
Showing 1- 7 of 7