Advantec Grade No. 235 Filter Paper for Very Fine Retention
Manufactured to exacting standards for uniform performance
  • Highest retention efficiency, retains very fine particulates, very slow flow rate, smooth
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
3,400.00 - ₹11,400.00INR / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N02357.0CM Grade No. 235 Qualitative Filter Paper; 7.0 cm dia.; pack of 100 8105022 Mfr # N02357.0CM
COMPARE
7 Alpha Cotton Cellulose
₹3,400.00INR / Pkg of 100
Advantec N023511.0CM Grade No. 235 Qualitative Filter Paper; 11.0 cm dia.; pack of 100 8105026 Mfr # N023511.0CM
COMPARE
11 Alpha Cotton Cellulose
₹4,600.00INR / Pkg of 100
Advantec N023512 5CM Grade No. 235 Qualitative Filter Paper; 12.5 cm dia.; pack of 100 8105028 Mfr # N023512 5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹5,500.00INR / Pkg of 100
Advantec N023515.0CM Grade No. 235 Qualitative Filter Paper; 15.0 cm dia.; pack of 100 8105030 Mfr # N023515.0CM
COMPARE
15 Alpha Cotton Cellulose
₹7,300.00INR / Pkg of 100
Advantec N023518.5CM Grade No. 235 Qualitative Filter Paper; 18.5 cm dia.; pack of 100 8105032 Mfr # N023518.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹11,400.00INR / Pkg of 100