Advantec Grade No. 232 Filter Paper for Medium / Medium-Fine Retention
Manufactured to exacting standards for uniform performance
  • Retains medium to medium-fine particulates, slow flow rate, smooth, normal hardness
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
2,000.00 - ₹3,100.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N02325.5CM Grade No. 232 Qualitative Filter Paper; 5.5 cm dia.; pack of 100 8104884 Mfr # N02325.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹2,000.00INR / Pkg of 100
Advantec N023211.0CM Grade No. 232 Qualitative Filter Paper; 11.0 cm dia.; pack of 100 8104890 Mfr # N023211.0CM
COMPARE
11 Alpha Cotton Cellulose
₹3,100.00INR / Pkg of 100