Advantec Grade No. 231 Filter Paper for Medium Retention
Manufactured to exacting standards for uniform performance
  • Medium retention; retains crystalline precipitates, moderate flow rate, smooth surface, normal hardness
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
1,900.00 - ₹6,500.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N02317.0CM Grade No. 231 Qualitative Filter Paper; 7.0 cm dia.; pack of 100 8104866 Mfr # N02317.0CM
COMPARE
7 Alpha Cotton Cellulose
₹1,900.00INR / Pkg of 100
Advantec N023111.0CM Grade No. 231 Qualitative Filter Paper; 11.0 cm dia.; pack of 100 8104870 Mfr # N023111.0CM
COMPARE
11 Alpha Cotton Cellulose
₹3,800.00INR / Pkg of 100
Advantec N023115.0CM Grade No. 231 Qualitative Filter Paper; 15.0 cm dia.; pack of 100 8104874 Mfr # N023115.0CM
COMPARE
15 Alpha Cotton Cellulose
₹6,500.00INR / Pkg of 100
Advantec N023118.5CM Grade No. 231 Qualitative Filter Paper; 18.5 cm dia.; pack of 100 8104880 Mfr # N023118.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹5,400.00INR / Pkg of 100