Advantec Grade No. 2 Filter Paper for Medium Retention
Manufactured to exacting standards for uniform performance
  • Medium retention
  • Retains medium crystalline precipitates
  • Fast flow rate
  • Smooth surface
  • Normal hardness
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
1,400.00 - ₹14,000.00INR / Pkg of 100
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec Grade No. 2 Qualitative Filter Paper, 5.5 cm Diameter; Pack of 100 8104834 Mfr # N025.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹1,400.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 7.0 cm Diameter; Pack of 100 8104836 Mfr # N027.0CM
COMPARE
7 Alpha Cotton Cellulose
₹1,700.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 9.0 cm Diameter; Pack of 100 8104838 Mfr # N029.0CM
COMPARE
9 Alpha Cotton Cellulose
₹2,000.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 11.0 cm Diameter; Pack of 100 8104840 Mfr # N0211.0CM
COMPARE
11 Alpha Cotton Cellulose
₹2,400.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 12.5 cm Diameter; Pack of 100 8104842 Mfr # N0212.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹2,500.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 15.0 cm Diameter; Pack of 100 8104846 Mfr # N0215.0CM
COMPARE
15 Alpha Cotton Cellulose
₹3,600.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 18.5 cm Diameter; Pack of 100 8104848 Mfr # N0218.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹5,400.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 32.0 cm Diameter; Pack of 100 8104854 Mfr # N0232.0CM
COMPARE
32 Alpha Cotton Cellulose
₹14,000.00INR / Pkg of 100
Advantec Grade No. 2 Qualitative Filter Paper, 4.7 cm Diameter; Pack of 100 8104858 Mfr # N024.7CM
COMPARE
4.7 Alpha Cotton Cellulose
₹1,400.00INR / Pkg of 100
Showing 1- 9 of 9