Advantec Grade No. 131 Filter Paper for Medium-Fine Retention
Manufactured to exacting standards for uniform performance
  • High retention efficiency for medium-fine crystalline precipitates like barium sulfate, slow flow rate, smooth surface, normal hardness
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
3,000.00 - ₹8,500.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N01315.5CM Grade No. 131 Qualitative Filter Paper; 5.5 cm dia.; pack of 100 8105004 Mfr # N01315.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹3,000.00INR / Pkg of 100
Advantec N013112.5CM Grade No. 131 Qualitative Filter Paper; 12.5 cm dia.; pack of 100 8105016 Mfr # N013112.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹5,900.00INR / Pkg of 100
Advantec N013115.0CM Grade No. 131 Qualitative Filter Paper; 15.0 cm dia.; pack of 100 8105018 Mfr # N013115.0CM
COMPARE
15 Alpha Cotton Cellulose
₹8,500.00INR / Pkg of 100
Advantec N013118.5CM Grade No. 131 Qualitative Filter Paper; 18.5 cm dia.; pack of 100 8105020 Mfr # N013118.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹6,800.00INR / Pkg of 100