Advantec Grade No. 101 Filter Paper for Coarse / Gelatinous Retention
Manufactured to exacting standards for uniform performance
  • Coarse retention; seed germination, retains large particles
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
1,300.00 - ₹8,800.00INR / Pkg of 100
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N01015.5CM Grade No. 101 Qualitative Filter Paper; 5.5 cm dia.; pack of 100 8105036 Mfr # N01015.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹2,200.00INR / Pkg of 100
Advantec N01017.0CM Grade No. 101 Qualitative Filter Paper; 7.0 cm dia.; pack of 100 8105038 Mfr # N01017.0CM
COMPARE
7 Alpha Cotton Cellulose
₹1,300.00INR / Pkg of 100
Advantec N01019.0CM Grade No. 101 Qualitative Filter Paper; 9.0 cm dia.; pack of 100 8105040 Mfr # N01019.0CM
COMPARE
9 Alpha Cotton Cellulose
₹2,900.00INR / Pkg of 100
Advantec N010112.5CM Grade No. 101 Qualitative Filter Paper; 12.5 cm dia.; pack of 100 8105050 Mfr # N010112.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹4,600.00INR / Pkg of 100
Advantec N010115.0CM Grade No. 101 Qualitative Filter Paper; 15.0 cm dia.; pack of 100 8105052 Mfr # N010115.0CM
COMPARE
15 Alpha Cotton Cellulose
₹6,200.00INR / Pkg of 100
Advantec N010118.5CM Grade No. 101 Qualitative Filter Paper; 18.5 cm dia.; pack of 100 8105054 Mfr # N010118.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹8,800.00INR / Pkg of 100