Advantec Grade No. 1 Filter Paper for Coarse Retention
Manufactured to exacting standards for uniform performance
  • Coarse retention; retains large crystalline particles and gelatinous precipitates. Fast flow rate, smooth surface, normal hardness.
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248°F (120°C). Sold in packs of 100.
1,200.00 - ₹5,000.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N015.5CM Grade No. 1 Qualitative Filter Paper; 5.5 cm dia.; pack of 100 8104804 Mfr # N015.5CM
COMPARE
5.5 Alpha Cotton Cellulose
₹1,200.00INR / Pkg of 100
Advantec N017.0CM Grade No. 1 Qualitative Filter Paper; 7.0 cm dia.; pack of 100 8104806 Mfr # N017.0CM
COMPARE
7 Alpha Cotton Cellulose
₹1,200.00INR / Pkg of 100
Advantec N019.0CM Grade No. 1 Qualitative Filter Paper; 9.0 cm dia.; pack of 100 8104808 Mfr # N019.0CM
COMPARE
9 Alpha Cotton Cellulose
₹1,800.00INR / Pkg of 100
Advantec N0111.0CM Grade No. 1 Qualitative Filter Paper; 11.0 cm dia.; pack of 100 8104810 Mfr # N0111.0CM
COMPARE
11 Alpha Cotton Cellulose
₹2,300.00INR / Pkg of 100
Advantec N0112.5CM Grade No. 1 Qualitative Filter Paper; 12.5 cm dia.; pack of 100 8104812 Mfr # N0112.5CM
COMPARE
12.5 Alpha Cotton Cellulose
₹2,400.00INR / Pkg of 100
Advantec N0115.0CM Grade No. 1 Qualitative Filter Paper; 15.0 cm dia.; pack of 100 8104814 Mfr # N0115.0CM
COMPARE
15 Alpha Cotton Cellulose
₹3,400.00INR / Pkg of 100
Advantec N0118.5CM Grade No. 1 Qualitative Filter Paper; 18.5 cm dia.; pack of 100 8104816 Mfr # N0118.5CM
COMPARE
18.5 Alpha Cotton Cellulose
₹5,000.00INR / Pkg of 100
Showing 1- 7 of 7