Advantec Grade GD120 Glass Fiber Filters
For analytical or gravimetric studies
Membrane prefilter (<1.2 µm), high wet strength and high loading capacity.
3,100.00 - ₹70,000.00INR / Box of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material
Availability Pricing
Advantec GD12047MM Grade GD120 Glass Fiber Filters; 47mm dia; 50/box 0664632 Mfr # GD12047MM
COMPARE
47 Borosilicate Glass Microfiber
₹3,100.00INR / Box of 50
Advantec GD12090MM Grade GD120 Glass Fiber Filters; 90mm dia; 50/box 0664634 Mfr # GD12090MM
COMPARE
90 Borosilicate Glass Microfiber
₹7,800.00INR / Box of 50
Advantec GD120125MM Grade GD120 Glass Fiber Filters; 125mm dia; 50/box 0664635 Mfr # GD120125MM
COMPARE
125 Borosilicate Glass Microfiber
₹16,000.00INR / Box of 50
Advantec GD120293MM Grade GD120 Glass Fiber Filters; 293mm dia; 50/box 0665613 Mfr # GD120293MM
COMPARE
293 Glass Microfiber
₹70,000.00INR / Pkg of 50
Advantec GD12025MM Grade GD120 Glass Fiber Filters; 25mm dia; 50/box 0665730 Mfr # GD12025MM
COMPARE
25 Glass Microfiber
₹4,000.00INR / Pkg of 50