Advantec Grade GB140 Glass Fiber Filters
For analytical or gravimetric studies
Low absorption filters. Slower filtration for industrial waste analysis.
9,300.00 - ₹55,400.00INR / Box of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (micron)
Availability Pricing
Advantec GB14047MM Grade GB140 Glass Fiber Filters; 47mm dia; 100/box 0664612 Mfr # GB14047MM
COMPARE
47 Borosilicate Glass Microfiber 0.4
₹14,500.00INR / Box of 100
Advantec GB14090MM Grade GB140 Glass Fiber Filters; 90mm dia; 100/box 0664614 Mfr # GB14090MM
COMPARE
90 Borosilicate Glass Microfiber 0.4
₹33,800.00INR / Box of 100
Advantec GB140125MM Grade GB140 Glass Fiber Filters; 125mm dia; 100/box 0664615 Mfr # GB140125MM
COMPARE
125 Borosilicate Glass Microfiber 0.4
₹55,400.00INR / Box of 100
Advantec GB14025MM Grade GB140 Glass Fiber Filters; 25mm dia; 100/box 0664684 Mfr # GB14025MM
COMPARE
25 Glass Microfiber 0.4
₹9,300.00INR / Pkg of 100