Advantec Grade GA55 Glass Fiber Filters
For analytical or gravimetric studies
General purpose; widely used in air pollution monitoring.
5,500.00 - ₹27,000.00INR / Box of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (micron)
Availability Pricing
Advantec GA5547MM Grade GA55 Glass Fiber Filters, 47mm dia; 100/box 0664602 Mfr # GA5547MM
COMPARE
47 Borosilicate Glass Microfiber 0.6
₹7,400.00INR / Box of 100
Advantec GA5590MM Grade GA55 Glass Fiber Filters; 100/box 0664604 Mfr # GA5590MM
COMPARE
90 Borosilicate Glass Microfiber 0.6
₹14,800.00INR / Box of 100
Advantec GA55125MM Grade GA55 Glass Fiber Filters, 125mm dia; 100/box 0664605 Mfr # GA55125MM
COMPARE
125 Borosilicate Glass Microfiber 0.6
₹27,000.00INR / Box of 100
Advantec GA5525MM Grade GA55 Glass Fiber Filters, 25mm dia; 100/box 0665700 Mfr # GA5525MM
COMPARE
25 Glass Microfiber 0.6
₹5,500.00INR / Pkg of 100