Advantec Extraction Thimbles, Quartz Fiber
Seamless, high-purity filters for extraction and air monitoring
  • Collection efficiency is 99.95%
Thimble filters are used in soxhlet apparatus for dust sampling and for extracting soluble components using organic solvents.
Quartz Fiber Thimbles are tapered for ease in loading into stack samplers. Heat resistant up to 1832°F (1000°C). Thickness is 2.2 mm.
16,700.00 - ₹36,600.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Height (mm) Material
Availability Pricing
Advantec N088R30X100MM Extraction Thimbles, Quartz Fiber, 30 mm ID x 100 mm H; 25/pack 2982134 Mfr # N088R30X34X100MM
COMPARE
30 100 Quartz Microfiber
₹36,600.00INR / Pkg of 25
Advantec N088RH25X90MM Extraction Thimbles, Quartz (Silica) Fiber, 25 mm ID x 90 mm H; 10/Pk 2982142 Mfr # N088RH25X90MM
COMPARE
25 Quartz (Silica)
₹16,700.00INR / Pkg of 10