Advantec Extraction Thimbles, Glass Fiber
Seamless, high-purity filters for extraction and air monitoring
  • Collection efficiency is 99.95%
Thimble filters are used in soxhlet apparatus for dust sampling and for extracting soluble components using organic solvents.
Glass fiber thimbles of borosilicate glass are acid washed for minimum trace metal content. They are acid resistant (except hydrofluoric) and ideal for high temperature extractions up to 932°F (500°C). Thickness is 1.6 mm.
24,700.00 - ₹36,600.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Height (mm) Material
Availability Pricing
Advantec N086R30X100MM Extraction Thimbles, Glass Fiber, 30 mm ID x 100 mm H; 25/pack 2982122 Mfr # N086R30X34X100MM
COMPARE
30 100 Glass Fiber
₹24,700.00INR / Pkg of 25
Advantec N088R30X80MM 88R Extraction Thimbles, Glass Fiber, 30 mm ID x 80 mm H; 25/pack 2982136 Mfr # N088R30X34X80MM
COMPARE
30 80 Quartz Microfiber
₹36,600.00INR / Pkg of 25