Advantec Disposable Syringe Filters, PTFE Hydrophobic
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
PTFE Hydrophobic Filterswith a laminated polypropylene support can be used to sterilize and clarify strong acids and solvents. Also typically used in gas filtration or venting applications.
22,600.00 - ₹38,000.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
Advantec 03JP050AN Syringe Filters, 0.5 um Hydrophobic PTFE, 3 mm; 100/Pk 8105402 Mfr # 03JP050AN
COMPARE
3 0.5 No
₹22,600.00INR / Pkg of 100
Advantec 13JP020AN Syringe Filters, 0.2 um Hydrophobic PTFE, 13 mm; 100/Pk 8105416 Mfr # 13JP020AN
COMPARE
13 0.2 No
₹34,650.00INR / Pkg of 100
Advantec 13JP050AN Syringe Filters, 0.5 um Hydrophobic PTFE, 13 mm; 100/Pk 8105418 Mfr # 13JP050AN
COMPARE
13 0.5 No
₹34,300.00INR / Pkg of 100
Advantec 25JP020AN Syringe Filters, 0.2 um Hydrophobic PTFE, 25 mm; 50/Pk 8105446 Mfr # 25JP020AN
COMPARE
25 0.2 No
₹38,000.00INR / Pkg of 50
Advantec 25JP050AN Syringe Filters, 0.5 um Hydrophobic PTFE, 25 mm; 50/Pk 8105448 Mfr # 25JP050AN
COMPARE
25 0.5 No
₹37,300.00INR / Pkg of 50
Advantec 50JP020AN Syringe Filters, 0.2 um Hydrophobic PTFE, 50 mm; 10/Pk 8105468 Mfr # 50JP020AN
COMPARE
50 0.2 No
₹30,700.00INR / Pkg of 10
Advantec 50JP050AN Syringe Filters, 0.5 um Hydrophobic PTFE, 50 mm; 10/Pk 8105478 Mfr # 50JP050AN
COMPARE
50 0.5 No
₹30,700.00INR / Pkg of 10
Showing 1- 7 of 7