Advantec Disposable Syringe Filters, Cellulose Acetate
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
Cellulose Acetate Filters feature hydrophilic membranes with low protein binding, suitable for aqueous protein solutions. The filters are nitrate-free, making them suitable for groundwater filtration.
19,800.00 - ₹39,600.00INR / Pkg of 50
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
Advantec 03CP020AS Syringe Filters, 0.2 um CA, 3 mm, Sterile; 100/Pk 8105400 Mfr # 03CP020AS
COMPARE
3 0.2 Yes
₹26,900.00INR / Pkg of 100
Advantec 13CP020AN Syringe Filters, 0.2 um CA, 13 mm; 100/Pk 8105406 Mfr # 13CP020AN
COMPARE
13 0.2 No
₹34,400.00INR / Pkg of 100
Advantec 13CP020AS Syringe Filters, 0.2 um CA, 13 mm, Sterile; 100/Pk 8105408 Mfr # 13CP020AS
COMPARE
13 0.2 Yes
₹38,900.00INR / Pkg of 100
Advantec 25CS020AN Syringe Filters, 0.20 um CA, 25 mm; 50/Pk 8105430 Mfr # 25CS020AN
COMPARE
25 0.2 No
₹20,300.00INR / Pkg of 50
Advantec 25CS020AS Syringe Filters, 0.20 um CA, 25 mm, Sterile; 50/Pk 8105432 Mfr # 25CS020AS
COMPARE
25 0.2 Yes
₹25,300.00INR / Pkg of 50
Advantec 25CS045AN Syringe Filters, 0.45 um CA, 25 mm; 50/Pk 8105434 Mfr # 25CS045AN
COMPARE
25 0.45 No
₹20,300.00INR / Pkg of 50
Advantec 25CS045AS Syringe Filters, 0.45 um CA, 25 mm, Sterile; 50/Pk 8105436 Mfr # 25CS045AS
COMPARE
25 0.45 Yes
₹25,300.00INR / Pkg of 50
Advantec 25CS080AN Syringe Filters, 0.80 um CA, 25 mm; 50/Pk 8105438 Mfr # 25CS080AN
COMPARE
25 0.8 No
₹19,800.00INR / Pkg of 50
Advantec 25CS080AS Syringe Filters, 0.80 um CA, 25 mm, Sterile; 50/Pk 8105440 Mfr # 25CS080AS
COMPARE
25 0.8 Yes
₹20,300.00INR / Pkg of 50
Advantec 50CP020AN Syringe Filters, 0.20 um CA, 50 mm; 10/Pk 8105460 Mfr # 50CP020AN
COMPARE
50 0.2 No
₹23,000.00INR / Pkg of 10
Advantec 50CP020AS Syringe Filters, 0.20 um CA, 50 mm, Sterile; 10/Pk 8105462 Mfr # 50CP020AS
COMPARE
50 0.2 Yes
₹25,900.00INR / Pkg of 10
Advantec 50CP045AN Syringe Filters, 0.45 um CA, 50 mm; 10/Pk 8105464 Mfr # 50CP045AN
COMPARE
50 0.45 No
₹23,000.00INR / Pkg of 10
Advantec 50CP045AN Syringe Filters, 0.45 um CA, 50 mm, Sterile; 10/Pk 8105466 Mfr # 50CP045AN
COMPARE
50 0.45 Yes
₹22,600.00INR / Pkg of 10
Advantec 13CP045AN Syringe Filters, 0.45 um CA, 13 mm; 100/Pk 8105474 Mfr # 13CP045AN
COMPARE
13 0.45 No
₹39,600.00INR / Pkg of 100
Showing 1- 14 of 14