Advantec Disposable Sterile Syringe Filters, PES
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
Polyethersolfone (PES) Filters are hydrophilic and low protein-binding; ideal for pharmaceutical and biological applications where sample recovery and low extractables are essential. Also used for filtration of high-contaminant or viscous liquids.
26,600.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
Advantec 25SS020RS Disposable Syringe Filters, PES; 0.2 µm; 25 mm dia; sterile; 50/PK WW-81054-92 Mfr # 25SS020RS
COMPARE
25 0.2 Yes
₹26,600.00INR / Pkg of 50
Advantec 25SS045RS Disposable Syringe Filters, PES; 0.45 µm; 25 mm dia; sterile; 50/PK WW-81054-94 Mfr # 25SS045RS
COMPARE
25 0.45 Yes
₹26,600.00INR / Pkg of 50