Advantec Disposable 50-mm Sterile Petri Dishes
No need to presterilize
  • Sterilized dishes are ready to use
  • Use only what you need—come in convenient packs of 20
  • Polystyrene dishes come with or without 47-mm absorbent cellulose pad that holds 1.8 to 2.0 mL of liquid media
  • Ridges for easy stacking
Ideal for culturing microorganisms in water/wastewater testing
21,400.00 - ₹67,600.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Outside Diameter (OD) (cm) Description
Availability Pricing
Advantec 800101 Petri Dish with Cellulose Pads, 50 mm dia x 11 mm H, 100/pk 1400520 Mfr # 42004A01
COMPARE
4.9375 Petri Dish with Cellulose Pads, 50 mm dia x 11 mm H, 100/pk
₹21,400.00INR / Pkg of 100
Advantec Petri dish without pads, 50x11 mm, 500/cs 1400526 Mfr # 42004A02
COMPARE
5 Petri dish without pads, 50x11 mm, 500/cs
₹59,500.00INR / Pkg of 500
Advantec 800501 Petri Dish with Cellulose Pads, 50 mm dia x 11 mm H, 500/pk 1400528 Mfr # 42004A03
COMPARE
5 Petri Dish with Cellulose Pads, 50 mm dia x 11 mm H, 500/pk
₹67,600.00INR / Pkg of 500