Advantec Cellulose Acetate Membranes
Features lower binding affinities for charged macromolecules
  • Membranes withstand temperatures to 180°C (356°F)
Cellulose acetate membranes have lower binding affinities for charged macromolecules then cellulose nitrate. Retention is primarily based on precipitate size — use to separate bound and unbound proteins. Membranes also have low-swelling properties making them ideal for use with alcohol based solutions.
29,600.00 - ₹120,000.00INR / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Advantec C020A025A Cellulose Acetate Membranes; 25 mm dia; 0.20 µm; 100/pack 0664502 Mfr # C020A025A
COMPARE
25 Cellulose Acetate 0.2
₹29,600.00INR / Pkg of 100
Advantec C020A047A Cellulose Acetate Membranes; 47 mm dia; 0.20 µm; 100/pack 0664504 Mfr # C020A047A
COMPARE
47 Cellulose Acetate 0.2
₹37,800.00INR / Pkg of 100
Advantec C020A090C Cellulose Acetate Membranes; 90 mm dia; 0.20 µm; 25/pack 0664506 Mfr # C020A090C
COMPARE
90 Cellulose Acetate 0.2
₹30,200.00INR / Pkg of 25
Advantec C020A142C Cellulose Acetate Membranes; 142 mm dia; 0.20 µm; 25/pack 0664508 Mfr # C020A142C
COMPARE
142 Cellulose Acetate 0.2
₹53,600.00INR / Pkg of 25
Advantec C045A025A Cellulose Acetate Membranes; 25 mm dia; 0.45 µm; 100/pack 0664512 Mfr # C045A025A
COMPARE
25 Cellulose Acetate 0.45
₹29,600.00INR / Pkg of 100
Advantec C045A047A Cellulose Acetate Membranes; 47 mm dia; 0.45 µm; 100/pack 0664514 Mfr # C045A047A
COMPARE
47 Cellulose Acetate 0.45
₹37,800.00INR / Pkg of 100
Advantec C045A090C Cellulose Acetate Membranes; 90 mm dia; 0.45 µm; 25/pack 0664516 Mfr # C045A090C
COMPARE
90 Cellulose Acetate 0.45
₹30,200.00INR / Pkg of 25
Advantec C045A142C Cellulose Acetate Membranes; 142 mm dia; 0.45 µm; 25/pack 0664518 Mfr # C045A142C
COMPARE
142 Cellulose Acetate 0.45
₹53,600.00INR / Pkg of 25
Advantec C080A025A Cellulose Acetate Membranes; 25 mm dia; 0.8 µm; 100/pack 0664522 Mfr # C080A025A
COMPARE
25 Cellulose Acetate 0.8
₹29,600.00INR / Pkg of 100
Advantec C080A047A Cellulose Acetate Membranes; 47 mm dia; 0.8 µm; 100/pack 0664524 Mfr # C080A047A
COMPARE
47 Cellulose Acetate 0.8
₹37,800.00INR / Pkg of 100
Advantec C080A090C Cellulose Acetate Membranes; 90 mm dia; 0.8 µm; 25/pack 0664526 Mfr # C080A090C
COMPARE
90 Cellulose Acetate 0.8
₹30,200.00INR / Pkg of 25
Advantec C020A037A Cellulose Acetate Membranes, 0.20 37mm Wp 100/pk 3623032 Mfr # C020A037A
COMPARE
37 Polycarbonate 0.2
₹36,800.00INR / Pkg of 100
Advantec C020A293C Cellulose Acetate Membranes; 293 mm dia; 0.20 µm; 25/pack 3623034 Mfr # C020A293C
COMPARE
293 Cellulose Acetate 0.2
₹120,000.00INR / Pkg of 25
Advantec C300A025A Cellulose Acetate Membranes; 25 mm dia, 3.0 µm pore; 100/pk 3623072 Mfr # C300A025A
COMPARE
25 Cellulose Acetate 3
₹29,600.00INR / Pkg of 100
Advantec C300A047A Cellulose Acetate Membranes; 47 mm dia, 3.0 µm pore; 100/pk 3623074 Mfr # C300A047A
COMPARE
47 Cellulose Acetate 3
₹37,800.00INR / Pkg of 100
Showing 1- 15 of 15