Advantec Blotting and Chromatography Paper

High quality papers for electrophoresis and chromatography applications
These papers are carefully tested for spot formation, capillary action, water flow rate, and absorption speed to ensure performance uniformity and repeatability. Applications include chromatography, electrophoresis/blotting, and separation of heavily loaded solutes.
9,000.00 - ₹144,000.00INR / Box of 100

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec N0514A460X570MM Paper, blotting and chromatography; Grade 514A, 460 x 570 mm; pack of 100 sheets 0664584 Mfr # N0514A460X570MM
COMPARE
Paper, blotting and chromatography; Grade 514A, 460 x 570 mm; pack of 100 sheets
₹144,000.00INR / Box of 100
Advantec N051A200X200MM Paper, blotting and chromatography; Grade 51A, 200 x 200 mm; pack of 100 sheets 2850170 Mfr # N051A200X200MM
COMPARE
Paper, blotting and chromatography; Grade 51A, 200 x 200 mm; pack of 100 sheets
₹15,400.00INR / Pkg of 100
Advantec N051B200X200MM Paper, blotting and chromatography; Grade 51B, 200 x 200 mm; pack of 100 sheets 2850171 Mfr # N051B200X200MM
COMPARE
Paper, blotting and chromatography; Grade 51B, 200 x 200 mm; pack of 100 sheets
₹9,800.00INR / Pkg of 100
Advantec N051B460X570MM Paper, blotting and chromatography; Grade 51B, 460 x 570 mm; pack of 100 sheets 2850172 Mfr # N051B460X570MM
COMPARE
Paper, blotting and chromatography; Grade 51B, 460 x 570 mm; pack of 100 sheets
₹41,200.00INR / Pkg of 100
Advantec N0514A200X200MM Paper, blotting and chromatography; Grade 514A, 200 x 200 mm; pack of 100 sheets 2850173 Mfr # N0514A200X200MM
COMPARE
Paper, blotting and chromatography; Grade 514A, 200 x 200 mm; pack of 100 sheets
₹9,000.00INR / Pkg of 100
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat