Advantec Aseptic Polysulfone Filter Systems
Reusable and versatile
  • Recessed filter support prevents twisting or tearing of the membrane
  • Holders accept 47-mm filters
These 47-mm filter holders feature recessed filter support that provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured. Each unit includes a graduated funnel (either 300-mL standard or 500-mL wide-mouth), silicone rubber stopper and O-ring, and a base. The #8B stopper adapts to standard 1-L flasks. Models 06644-72 and -76 have a funnel cover that can be vented aseptically using a disposable syringe filter (included). Model 06644-76 has a 300-mL receiver interchangeable with all other models.
10,700.00 - ₹16,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Funnel Size (mL) Description
Availability Pricing
Advantec 43301030 Aseptic Polysulfone Filter Holder, 300 mL, with Funnel and Base WW-06644-70 Mfr # 43301030
COMPARE
300 Aseptic Polysulfone Filter Holder, 300 mL, with Funnel and Base
₹10,700.00INR / Each
Advantec 43301020 Aseptic Polysulfone Filter Unit, 300 mL, with Funnel, Base, and Cover WW-06644-72 Mfr # 43301020
COMPARE
300 Aseptic Polysulfone Filter Unit, 300 mL, with Funnel, Base, and Cover
₹11,600.00INR / Each
Advantec 43301050 Wide-Mouth Polysulfone Filter Holder, 500 mL Funnel and Base WW-06644-74 Mfr # 43301050
COMPARE
500 Wide-Mouth Polysulfone Filter Holder, 500 mL Funnel and Base
₹13,500.00INR / Each
Advantec 43301010 Complete Aseptic Polysulfone Filter System, 300 mL WW-06644-76 Mfr # 43301010
COMPARE
300 Complete Aseptic Polysulfone Filter System, 300 mL
₹16,000.00INR / Each