Adam Equipment IHS Crane Scales
Solid construction for suspended weighing
  • Selectable readability for different environments
  • Large bright red LED display is easy to read in any lighting condition
  • Rechargeable battery for up to 150 hours of usage
  • Remote control for easy operation
  • Robust metal die cast housing
  • 360° rotational hook
The IHS crane scales offer durable easy-to-use weighing with smart features that take the hassle out of suspension weighing. With a range of capacities to suite different needs the IHS crane scales works for many different applications.
A standard remote control makes for safe and simple use and the large red display is easy to view in any lighting condition. A solid construction with metal housing and 360° rotational hook offers complete flexibility and ease of use.
Internal rechargeable battery with over 150 hours of use lets you position this crane scale where you need it. Just charge it up and use it when you need to.
Display hold function lets you hold the value on the display which can be view once the weight has been removed.
For easy operation the remote control allows you to operate the crane scale without needing to be near the unit. All the basic function of tare and holding values are
accessible.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment Battery/AC Operated Crane Scales; 1000kg x 0.2kg; 120 V 1171015 Mfr # IHS 2A
COMPARE
1000 120 200
Call For Price
Adam Equipment Battery/AC Operated Crane Scales; 3000kg x 0.5kg; 120 V 1171016 Mfr # IHS 6A
COMPARE
3000 120 500
Call For Price
Adam Equipment Battery/AC Operated Crane Scales; 5000kg x 1kg; 120 V 1171017 Mfr # IHS 10A
COMPARE
5000 120 1000
Call For Price
Adam Equipment Battery/AC Operated Crane Scales; 10000kg x 2kg 120 V 1171018 Mfr # IHS 20A
COMPARE
10000 120 2000