Adam Equipment Aqua Compact Washdown Scales
Designed to withstand harsh working environments
  • IP67 rated against water and dust
  • Sealed membrane keypad protects against dirt and spills
  • Overload protection prevents damage to internal components
  • Heavy-duty rubber feet ensure stability of unit
  • Auto power-off conserves battery life
The Aqua washdown scales are IP67 rated with a durable solid ABS housing and a removable stainless-steel weighing pan ideal for food production, catering, and industrial environments. Dual displays, one front facing and one rear facing, offer enhanced visibility. Check weighing is fast and simple with built-in LED indicators that clearly tell you if the load is under, over, and within acceptable limits. The parts counting optimization feature automatically refines piece weight as parts are added. Power is supplied by AC adapter or rechargeable battery.
85,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (kg) Readability (g)
Availability Pricing
Adam Equipment ABW 4 Aqua Compact Washdown Scales, 8.8lb/4kg x 0.001lb / 0.5g WW-20004-85 Mfr # ABW 4
COMPARE
External 32 5
₹85,600.00INR / Each
Adam Equipment ABW 8 Aqua Compact Washdown Scales, 18lb/8kg x 0.002lb / 1g WW-20004-86 Mfr # ABW 8
COMPARE
External 16 2
₹85,600.00INR / Each
Adam Equipment ABW 16 Aqua Compact Washdown Scales, 35lb/16kg x 0.005lb / 2g WW-20004-87 Mfr # ABW 16
COMPARE
External 8 1
₹85,600.00INR / Each
Adam Equipment ABW 32 Aqua Compact Washdown Scales, 70lb/32kg x 0.01lb / 5g WW-20004-88 Mfr # ABW 32
COMPARE
External 4 0.5
₹85,600.00INR / Each