Adam Equipment Anti-vibration Balance Table
Provides suitable working conditions for sensitive devices
  • Large working surface of 400mm x 450mm
  • Ergonomic Styling for user comfort
  • Rugged construction for durability
  • Easy to set up light weight aluminium frame
  • Adjustable levelling feet to take care of un-even floor
  • New aluminium tubular frame for light weight
  • Scratch Resistant granite slab
  • Polished finish
Anti-Vibration table is designed for use in the laboratory or metrology room to provide suitable working conditions for devices that are sensitive to vibrations and shocks.
290,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Length (in) Width (in)
Availability Pricing
Adam Equipment 104008036 Balance Antivibration Table 1112092 Mfr # 104008036
COMPARE
30 51/64 31 1/2
₹290,000.00INR / Each