Accident Prevention Tags; Danger: Do Not Operate - Maintenance Department
OSHA/ANSI-Compliant Header Tags meet OSHA 29 CFR 1910.145 (f) and ANSI Z535.5 standards which provide specifications, designs, and guidelines for the use and application of accident prevention tags. These tags clearly indicate a temporary hazard that may lead to personal injury or property damage; use until that hazard is eliminated or hazardous operation is complete.
All tags can be written on and have top gabled corners with 3⁄8" reinforced metal grommet on vinyl plastic.
6,700.00INR / Pkg of 25
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Legend Material
Availability Pricing
Accuform MDT206CTP Prevention Tags, Danger Do Not Operate, Back B, 5.8" x 3.1", Cardstock; 25/Pk 4050354 Mfr # MDT206CTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! Cardstock
₹6,700.00INR / Pkg of 25