3M BX Reader Safety Glasses
Three lens angles for a custom fit
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
High-impact polycarbonate lens blocks 99.9% UV and features antifog coating. Glasses feature a soft, universal fit nosebridge and soft temple provide comfort for long periods of wear. Temple is adjustable to three lengths. Reader magnifiers available in 1.5, 2.0, and 2.5 power
Compliance: ANSI Z87.1-2003, CSA Z94.3
7,200.00 - ₹10,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Color Magnification
Availability Pricing
3M 11374-00000 Eyewear, Silver/Black frame, Clear lens Reader, 1.5 Diopter 8647263 Mfr # 11374-00000-20
COMPARE
Black/Silver Clear +1.5
₹10,400.00INR / Each
3M 11376-00000 Eyewear, Silver/Black frame, Clear lens Reader, 2.5 Diopter 8647265 Mfr # 11376-00000-20
COMPARE
Black/Silver Clear +2.5
₹7,200.00INR / Each