3M Belt-Mounted powered Air Purifying Repirators (PAPR)
Provide high levels of airflow for extra comfort
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
These respirators contour to the hip and body for a comfortable fit. The waist belt and shoulder strap are durable and easy to decontaminate. Choose from a full-face, half-mask or general-purpose hood respirator. The full-face mask respirators easily converts using DIN adapter and DIN plugs. Masks are made of soft lightweight material in three sizes. The hood respirator gives you respiratory protection while providing a flow of air to help you stay cool. Hooded respirator is ideal for workers with facial hair who can not wear tight fitting mask. The Motor Blower assembly includes the motor blower unit, waist belt, battery pack, battery charger, power cord, and flow meter.
Compliance NIOSH/MSHA
2,600.00INR / Case of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
3M 6880 Din Plugs for 6000 Series Full Face mask to Use with GVP PAPR WW-40120-05 Mfr # 6880
COMPARE
Din Plugs for 6000 Series Full Face mask to Use with GVP PAPR
₹2,600.00INR / Case of 1