Zefon Coal Dust Sampling Cassettes, MSHA Mandated Sampling, 2/Pk
Representative image only.
Zefon Coal Dust Sampling Cassettes, MSHA Mandated Sampling, 2/Pk
Zefon | Item: 6006233
Ensure precise and compliant coal dust sampling with Zefon’s Preweighed Coal Dust Sampling Cassettes
₹9,100.00INR / Pkg of 2
₹9,100.00INR / Pkg of 2
Product Description
Key Features
  • 5-micron PVC filter with stainless steel supports for optimal performance
  • Weight of each filter capsule recorded to the nearest 0.001 mg, ensuring accuracy
  • Includes a mine data card and a pre-addressed mailing carton for easy submission to the Mine Safety and Health Administration (MSHA
More About this Item
Each cassette features a sealed outer plastic capsule with an inner pre-weighed filter capsule, designed to meet the strict standards of 30 CFR part 74 for coal mine dust sampling.
Coal Dust Sampling Cassettes are not returnable and Zefon's normal 30 day return policy does not apply to this product.
Specifications
  • MaterialPVC filter with stainless steel supports
  • Pore Size (µm)5
  • DescriptionCoal Dust Sampling Cassettes, MSHA Mandated Sampling, 2/Pk