YSI 3682 Zobell Solution for ORP Calibration, 1.25 g

Representative image only.

YSI 3682 Zobell Solution for ORP Calibration, 1.25 g

YSI | Mfr: 061320| Item: 0547860

Add distilled water for fresh solution

₹16,000.00INR / Each

₹16,000.00INR / Each

Buy It Together

Current Item

YSI 3682 Zobell Solution for ORP Calibration, 1.25 g
YSI 3682 Zobell Solution for ORP Calibration, 1.25 g
0547860

₹16,000.00INR / Each
+ + + =
Total Price 88600.00 ADD 4 ITEMS TO CART
Product Description

Key Features

  • Reconstitute with 125 mL of distilled water for fresh solution
  • Guarantee freshness with write-on label
  • Redox potential value for Ag/AgCl electrodes is 231 ± 10 mV and 185 ± 10 mV for saturated calomel electrodes
  • 1 year warranty

More About this Item

Use Zobell solution to verify the performance of your ORP electrodes. Reagent is available in dry form and must be reconstituted with 125 mL of deionized or distilled water prior to use. Bottle label has a place to write the expiration date to guarantee freshness.

1 Year Warranty
Specifications
  • Volume1.25g
  • DescriptionZobell Solution for ORP Calibration, 1.25 g
  • Warranty1 year
Reviews
Q&A